Special marker pens
Catalog # GREI841077UK
Supplier: Greiner Bio-One
Specifications
- Sterile:-
- Colour:Black
- Description:Marker pen, Super fine
- Seq:02
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Marker pens designed for labelling all plastic articles and glassware.
- Water-resistant
- Quick drying
Ordering information: Available in four colours and two writing thicknesses.