Order Entry
Denmark
ContactUsLinkComponent
 
2,3-Dimethoxy-5-methyl-p-benzoquinone ≥97.0% (by titrimetric analysis)
 :  TCI
 
undefined
2,3-Dimethoxy-5-methyl-p-benzoquinone ≥97.0% (by titrimetric analysis)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Purity (Iodometric Titration):
    min. 97.0 %
  • Melting point:
    59.0 to 62.0 deg-C
  • Solubility in hot Water:
    almost transparency

 

Frequently Bought Together