Cyclohexyl phenyl ketone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:712-50-5
:
Formula:C₁₃H₁₆O
MDL Number:MFCD00001467
Boiling Pt:168°C
Molecular Weight:188.27 g/mol
EINECS:211-923-5
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:55.0 to 59.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together