1,2,4-Tribromobenzene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:615-54-3
:
Formula:C₆H₃Br₃
MDL Number:MFCD00000062
Molecular Weight:314.8 g/mol
Melting Pt:44
EINECS:210-433-9
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:40.0 to 44.0 deg-C
Specification Test Results
Frequently Bought Together