2,3-Dimethyl-1-butene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:563-78-0
:
Formula:C₆H₁₂
MDL Number:MFCD00008923
Boiling Pt:55°C
Molecular Weight:84.16 g/mol
Melting Pt:-157
Flash Pt:–18°C
Storage Temperature:Ambient
EINECS:209-262-2
UN:3295
ADR:3 II
:GHS02
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together