trans-4-Octene ≥99.0% (by GC)
: TCI
: (E)-4-Octene
:14850-23-8
:
:C₈H₁₆
:MFCD00009476
:122°C
:112.22 g/mol
:-93.8
:21°C
:Ambient
:238-913-3
:3295
:3 II
:GHS02
:GHS08
- Environmentally Preferable:
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Frequently Bought Together