n-Dodecane ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Dodecane
CAS Number:112-40-3
:
Formula:H₃C(CH₂)₁₀CH₃
MDL Number:MFCD00008969
Boiling Pt:216°C
Molecular Weight:170.34 g/mol
Melting Pt:-9.6
Flash Pt:82°C
EINECS:203-967-9
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together