Formamide
Supplier: G-Biosciences
Certificates
About this item
Synonyms: Carbamaldehyde
CAS Number:75-12-7
:
Formula:HCONH₂
MDL Number:MFCD00007941
Boiling Pt:210
Melting Pt:2
Density:1.1334 g/cm³ (20 °C)
Molecular Weight:45.04 g/mol
Flash Pt:79
Merck Index:13,04261
EINECS:200-842-0
:GHS08
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together