Indole-7-carboxylic acid ≥97.0% (by GC, titration analysis)
: TCI
: 1H-Indole-7-carboxylic acid
:1670-83-3
:
:C₉H₇NO₂
:MFCD00210442
:161.16 g/mol
:216-801-5
:GHS07
- Environmentally Preferable:
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:203.0 to 207.0 deg-C
Frequently Bought Together