Order Entry
Denmark
ContactUsLinkComponent
tert-Butanol ≥99.0% (by GC)
tert-Butanol ≥99.0% (by GC)
Supplier:  TCI
Certificates
undefined
tert-Butanol ≥99.0% (by GC)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
  • Purity (GC):
    min. 99.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together