trans-Stilbene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: trans-1,2-Diphenylethylene, (E)-1,2-Diphenylethene, trans-1,2-Diphenylethene
CAS Number:103-30-0
:
Formula:C₁₄H₁₂
MDL Number:MFCD00064300
Boiling Pt:307°C
Molecular Weight:180.25 g/mol
Merck Index:14,08817
EINECS:203-098-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 98.0 %
- Melting point:123.0 to 126.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together