Order Entry
Denmark
ContactUsLinkComponent
 
Plasmid Midiprep kit, GeneJET™
 
undefined
Plasmid Midiprep kit, GeneJET™

 

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Environmentally Preferable:

 

Frequently Bought Together