Myringotomy blade
Catalog # SWAN2102
Supplier: SWANN MORTON
Specifications
- Description:Myringotomy blades, packed in individual plastic trays
- Environmentally Preferable:
- Pk:10
Specifications
About this item
The sterile stainless steel Myringotomy blade is used in conjunction with the SF1 or SF13 Fine surgical handle in the treatment of 'Otitis media' or 'Glue ear'.
- Blade and handle made of stainless steel
- Blade is double egded so allowing for cutting movements in both directions
- Fits handles SF1, SF2, SF3, SF4, SF13 and SF23
The Myringotomy blade is used to make an incision in the tympanic membrane (ear drum) into which a 'grommet' can be placed to allow ventilation of the middle ear which has become blocked thus interfering with the patients ability to hear.