Adenosine-5'-triphosphate disodium salt (ATP disodium salt) trihydrate ≥99%, white powder cell culture reagent
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: (((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)triphosphoric acid, disodium salt trihydrate, 5'-ATP·2Na·3H2O, Adenosine-5'-triphosphoric acid disodium salt trihydrate, ATP, 2Na 3H2O, ATP disodium trihydrate
CAS Number:51963-61-2
:987-65-5
Formula:C₁₀H₁₄N₅Na₂O₁₃P₃·3H₂O
MDL Number:MFCD00150754
Molecular Weight:605.19 g/mol
Melting Pt:185…190 °C (lit.)
Storage Temperature:Freezer
EINECS:213-579-1
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test A:Passes
- Identity Test B:Passes
- Identity Test C:Passes
- Purity:~ 99%
- FTIR:Conforms to Standard
- I max (H20):258 ± 2 nm
- OD250/OD259:0.75-0.83
- pH (5% aq soln):Acidic to Litmus Paper
- Solubility (5% aq soln):Clear to Slightly Hazy Solution
Specification Test Results
Frequently Bought Together