Order Entry
Denmark
ContactUsLinkComponent
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate ≥96%, white powder
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate ≥96%, white powder
Supplier:  MP Biomedicals
Certificates
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate ≥96%, white powder
Supplier:  MP Biomedicals

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥96%
  • Moisture :
    Report Result
  • FTIR:
    Conforms to Standard
  • UV/Vis: l max (water):
    258 ± 2 nm
  • OD 250/OD 259:
    0.75 - 0.83
  • pH (5% aq soln):
    Acidic to Litmus Paper
  • Solubility (5% aq soln):
    Clear to Slightly Hazy Solution

Specification Test Results

Frequently Bought Together