Order Entry
Denmark
ContactUsLinkComponent
Streptavidin
Streptavidin
Catalog # MOLEM47454398
Supplier:  Molekula
CAS Number:  
Streptavidin
Catalog # MOLEM47454398
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 mg
  • CAS No.:
    9013-20-1
  • Source:
  • Species:
  • Environmentally Preferable:
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00082035
  • Protein/peptide name:
    Streptavidin

Specifications

About this item