Oil red O biological stain
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: CI 26125, Sudan Red 5B, Solvent Red 27
CAS Number:1320-06-5
:
Formula:C₂₆H₂₄N₄O
MDL Number:MFCD00003898
Molecular Weight:408.51 g/mol
Melting Pt:120 °C (dec.)
Storage Temperature:Ambient
EINECS:215-295-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Dark green to blue-red to brown to black Powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Dye content:≥70 %
Specification Test Results
Frequently Bought Together