Spiral plater, WASP 2
710-0915
- Description:Spiral plater WASP 2
- Environmentally Preferable:
- Pk:1
Spiral platers are used extensively in the food industry to test for safe levels of bacteria; non-food applications include pharmaceuticals, cosmetics, water, and mouth and skin bacteria. The WASP 2 (Whitley Automatic Spiral Plater) takes advantage of contemporary technology to offer microprocessor-controlled stepper motor motion to streamline and automate traditional spiral plate techniques.
- Eliminates the need for serial dilutions
- Requires minimal set up time
- Reduces cost per test
- Standardises counting methods
: Includes water pots, spare bleach bottle, disposable beakers and tool kit.