Betaine monohydrate 99+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:590-47-6
:
Formula:C₅H₁₁NO₂·1H₂O
MDL Number:MFCD00150010
Storage Temperature:Ambient
EINECS:203-490-6
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:11 % to 15 % (140°C, 3 h)
- Sulfated ash:≤0.1 %
- Solubility:(10% in H2O) Clear colorless solution
- Titration with HClO4:99.0 to 101.0 % (On dry substance)
- Chloride (Cl):≤200 ppm
- Sulfate (SO4):≤200 ppm
- Heavy metals (as Pb):≤10 ppm
Specification Test Results
Frequently Bought Together