Order Entry
Denmark
ContactUsLinkComponent
Stands for Imhoff sedimentation cones or seperatory funnels, SP Bel-Art
Stands for Imhoff sedimentation cones or seperatory funnels, SP Bel-Art
Catalog # BELAH189670000
undefined
Stands for Imhoff sedimentation cones or seperatory funnels, SP Bel-Art
Catalog # BELAH189670000
Specifications
  • For:
    eight cones or funnels
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
This high-density polyethylene rack holds four separatory funnels on the lower shelf or four long-stem funnels on the upper shelf [maximum funnel diameter 98 mm (3⅞")].
  • Set screws allow shelf height adjustments anywhere along the 74 cm (29") support rods
  • Includes 24 insert clips that allow the rack to accommodate different sized Separatory Funnels and Imhoff Cones
  • Chemical resistant and fully adjustable
Dimensions: 22×66×74 cm H (8,5×26×29")