4-Hydroxy-2(5H)-furanone 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Tetronic acid, beta-Tetronic acid, 2,4(3H,5H)-Furandione, Tetrahydrofuran-2,4-dione
CAS Number:541-57-1
:
Formula:C₄H₄O₃
MDL Number:MFCD00010086
Molecular Weight:100.07 g/mol
Melting Pt:∼ 140 °C (dec.)
EINECS:208-785-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together