Digital burettes, Titrette®
: Brand
Variable volume.
- Light, flexible burette with compact design and high accuracy
- Smooth running operation with large, easy grip hand wheels for sensitive, dropwise titration
- Easier use: Adjustment with easy calibration, calibration schedule to save a date for the next calibration, save power with Automatic Off function, change decimal place settings
- Easy to dismantle: Cleaning, maintenance and part replacement
- PC interface (optional) - for data transmission directly to a PC, which eliminates transmission errors during transcription of the primary data to fulfil important GLP requirements
Titrate quickly and reliably with high precision, even in close quarters, with no power connection needed.
The Titrette® has an innovative measuring system – tight error limits (relate to the nominal volume) of class A glass burettes as per DIN EN ISO 385 are met.
: Titrette® bottle-top burette, conformity certified, supplied with performance certificate, telescopic filling tube (170 to 330 mm), recirculation tube, two micro batteries (AAA/UM4/LR03), three bottle adapters PP (GL 45/32, GL 45/S 40 and GL 32/NS 29/32), two coloured light shield inspection windows and operating manual. Additional items supplied when ordering the Titrette® with PC interface: Connector cable - Sub-D plug connector, 9 pin - and a CD with driver software, communication protocol, example application and special operating manual.
- Catalog No:
- 613-0796
- 613-0906
- 613-0905
- 613-3840
- 613-0795
- 613-5796
- 613-2314
- Environmentally Preferable:
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Environmental certification:
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
- ISO 14001 and ISO 50001
Frequently Bought Together