About this item
The jetPRIME® DNA and/or siRNA transfection reagent is a powerful and versatile reagent designed to ensure high DNA transfection efficiency and excellent gene silencing in a variety of adherent cells.
- High DNA transfection efficiency
- Uses low amounts of nucleic acid
- Superior cell viability
- One reagent for DNA and/or siRNA transfection
- Cost effective
jetPRIME® is ideal for co-transfection of DNA and siRNA. jetPRIME® is gentle to cells since it requires low amounts of reagent and nucleic acid during transfection. Effective and non toxic DNA and siRNA delivery is essential for reliable scientific results. Free of charge samples are available upon request. Please contact your local Avantor sales office for further details.
Since 1st of September 2021 Polyplus-Tranfection changed product part numbers. Products are remaining the same and the change is only related to the product part number. New product part numbers have symbol 'New'. For duplicated products with symbol 'telephone' please be informed that it’s the same product but with previous part number which we will only offer till the end of stock.