Zinc(II) nitrate hexahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Zinc nitrate hexahydrate
CAS Number:10196-18-6
:
Formula:ZnN₂O₆·6H₂O
MDL Number:MFCD00149889
Boiling Pt:∼105 °C (1013 hPa)
Melting Pt:36
Density:2.06 g/cm³ (20 °C)
Molecular Weight:297.49 g/mol
Storage Temperature:Ambient
EINECS:231-943-8
ADR:5.1 II
Merck Index:12,10273
UN:1514
:GHS03
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together