Tubing
: S.C.A.T.
Flexible tubing made of PP and PTFE-EL material.
- Available in various materials, lengths and properties
Electrically conductive plastic tubing provides safe protection against ignition hazards and electrostatic charging.
Tubing cutter
Frequently Bought Together