Order Entry
Denmark
ContactUsLinkComponent
Sporicide, Minncare® Cold Sterilant
Sporicide, Minncare® Cold Sterilant
  148-0062
undefined
Sporicide, Minncare® Cold Sterilant
  148-0062
  • Description:
    Minncare® Cold Sterilant
  • Environmentally Preferable:
  • Packed:
    5 l
  • Pk:
    2
Minncare® Cold Sterilant concentrate is an effective viricide, bactericide, fungicide, and sporicide for surface disinfection in pharmaceutical cleanroom environments. It is completely and contains only pharmaceutical grade raw materials.
  • Leaves no measurable air residue following ventilation of room
  • Faster, safer, and less corrosive than aldehyde-based liquids
  • EPA registered for fogging as part of normal cleaning and disinfecting procedures