Silica gel, Pore size: 60 Å (0.015-0.040 mm) for column chromatography, Millipore®
1.15111.1000
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
- pH-value (10 % suspension):6.5 - 7.5
- Fe (Iron):≤ 0.02 %
- Loss on drying:≤ 9.0 %
- particle size (Malvern) (d10):11.0 - 17.0 µm
- particle size (Malvern) (d50):20.0 - 26.0 µm
- particle size (Malvern) (d90):34.0 - 40.0 µm
:
:60.08 g/mol
:2230
:1710
:Ambient
:231-545-4
:63231-67-4
:SiO₂
:MFCD00011232