Orthoboric acid 99.0-100.5% Ph. Eur.
: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
:10043-35-3
:
:B(OH)₃
:MFCD00011337
:185
:160
:1.44 g/cm³ (25 °C)
:61.83 g/mol
:13,01326
:Ambient
:233-139-2
:01-2119486683-25
:GHS08
- Environmentally Preferable:
- Assay:99.0 - 100.5 %
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes test
- Identification B:Passes test
- Appearance of solution:Passes test
- pH (3.3 %):3.8 - 4.8
- Solubility in alcohol:Passes test
- Organic matter:Passes test
- SO4 (Sulphate):Max. 450 ppm
- Heavy metals (as Pb):Max. 15 ppm
- Residual solvents:Unlikely by manuf.process
Frequently Bought Together