Orthoboric acid
Catalog # HACH1481799
Supplier: Hach
CAS Number:
Specifications
- Pk:100
- Seq:0100
- Environmentally Preferable:
Specifications
About this item
Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
:
:61.83 g/mol
Boiling Pt:185
EINECS:233-139-2
REACH:01-2119486683-25
:GHS08
Storage Temperature:Ambient
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337
Melting Pt:160
Density:1.44 g/cm³ (25 °C)
Merck Index:13,01326