Orthoboric acid 99+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
CAS Number:10043-35-3
:
Formula:B(OH)₃
MDL Number:MFCD00011337
Boiling Pt:185
Molecular Weight:61.83 g/mol
Melting Pt:169 °C (dec.)
Merck Index:13,01326
EINECS:233-139-2
REACH:01-2119486683-25
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder or crystals or granules
- Titration with NaOH:≥99.0 %
- Chloride (Cl):≤10 ppm
- Sulfate (SO4):≤145 ppm
- Iron (Fe):≤10 ppm
Specification Test Results
Frequently Bought Together