Tetrafluoroboric acid diethyl ether complex (50 - 55% HBF₄)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67969-82-8
:
Formula:C₄H₁₁BF₄O
MDL Number:MFCD00011345
Density:1.19
Molecular Weight:161.93 g/mol
Flash Pt:36 °C (97 °F)
Merck Index:14,4149
EINECS:267-983-8
UN:2920
:GHS05
:GHS02
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together