Sodium sulphite, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
: Merck
:7757-83-7
:
:Na₂SO₃
:MFCD00003503
:2.63 g/cm³ (20 °C)
:126.04 g/mol
:> 500 °C
:13,08757
:M-Clarity™ quality level = MQ300
:Ambient
:231-821-4
:01-2119537420-49
- Environmentally Preferable:
- Solubility:220 g/l (20 °C)
- Melting Point:500 °C (decomposition)
- Molar Mass:126.04 g/mol
- Bulk Density:1480 kg/m3
- Density:2.63 g/cm3 (20 °C)
- pH:8.8 - 10 (50 g/l, H2O, 20 °C)
Frequently Bought Together