Order Entry
Denmark
ContactUsLinkComponent
 
Lithium bromide
Catalog # 1.05669.1000
Supplier:  Merck
CAS Number:  
Lithium bromide
Catalog # 1.05669.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

Specifications

About this item

 :
 :86.84 g/mol
EINECS:231-439-8
 :GHS07
Storage Temperature:Ambient
CAS Number:7550-35-8
 :23303-71-1
Formula:LiBr
MDL Number:MFCD00011077
Boiling Pt:1265
Melting Pt:550
Density:3.46 g/cm³ (20 °C)
Merck Index:13,05548