1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
CAS Number:123-91-1
:
Formula:C₄H₈O₂
MDL Number:MFCD00006571
Boiling Pt:100…102 °C
Melting Pt:11.8
Density:1.034
Molecular Weight:88.11 g/mol
Flash Pt:12 °C (54 °F)
EINECS:204-661-8
UN:1165
ADR:3 II
Merck Index:14,3300
:Packaged under argon in resealable ChemSeal™ bottles.
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together