Ethyl-4-(bromomethyl)benzoate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ethyl 4-(bromomethyl)benzoate
CAS Number:26496-94-6
:
Formula:C₁₀H₁₁BrO₂
MDL Number:MFCD03791307
Boiling Pt:145 °C (5 mmHg)
Molecular Weight:243.1 g/mol
Melting Pt:28…33 °C
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together