Magnesium ethanolate (21 - 22% Mg)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Magnesium ethoxide, Magnesium ethylate
CAS Number:2414-98-4
:
Formula:C₄H₁₀MgO₂
MDL Number:MFCD00009060
Density:1.01
Molecular Weight:114.43 g/mol
Melting Pt:270 °C (dec.)
Storage Temperature:Ambient
EINECS:219-323-5
UN:3205
ADR:4.2 II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together