Benzyl benzoate 99+%
: Benzoic acid benzyl ester
:120-51-4
:
:C₁₄H₁₂O₂
:MFCD00003075
:323 °C (760 mmHg)
:18…20 °C
:1.118 g/cm³
:212.25 g/mol
:147 °C
:3082
:9 III
:13,01129
:204-402-9
:GHS07
:GHS09
:Ambient
- Environmentally Preferable:
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5670 to 1.5700 (20°C, 589 nm)
- GC:≥99.0 %
- Specific gravity:(20/20°C) 1.118 to 1.122
- Acid value:≤0.3 mg KOH/g
Frequently Bought Together