α'-Bromo-α,α,α-trifluoro-p-xylene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-(Bromomethyl)benzotrifluoride, 1-(Bromomethyl)-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)benzyl bromide
CAS Number:402-49-3
:
Formula:C₈H₆BrF₃
MDL Number:MFCD00000403
Boiling Pt:65…66 °C (5 mmHg)
Melting Pt:29…33 °C
Density:1.546
Molecular Weight:239.03 g/mol
Flash Pt:88 °C (190 °F)
Storage Temperature:Ambient
EINECS:206-947-8
UN:3261
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together