1-Phenyl-1-decanone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Decanophenone, Decyl phenyl ketone, Nonyl phenyl ketone
CAS Number:6048-82-4
:
Formula:C₆H₅C(O)(CH₂)₈CH₃
MDL Number:MFCD00009582
Boiling Pt:165…167 °C (7 mmHg)
Molecular Weight:232.37 g/mol
Melting Pt:34…36 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:227-946-9
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together