Order Entry
Denmark
ContactUsLinkComponent
 
2,6-Dimethoxyphenol 99%
 
undefined
2,6-Dimethoxyphenol 99%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to light yellow to grey to brown Crystalline powder or crystals or flakes or fused solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    52 to 57 °C
  • GC:
    ≥98.5 %

 

 

Frequently Bought Together