Plasma separator tubes, PST™ II, BD Vacutainer®
Supplier: BD Medical
Certificates
About this item
Plasma tubes with separating gel for clinical chemistry are available with spray-dried lithium heparin additives.
- It act as stable barrier between plasma and cells providing analyte stability and better sample quality
- Correct mixing of the tubes immediately after the blood sample collection is important to avoid micro clotting
- Plasma separation tubes should be gently inverted 180° and back 8 to 10 times
During the centrifugation of these tubes, an inert gel separates the plasma and the cells preventing the contamination of the plasma from the separated cellular components.
Ordering information: Study documentation in relation to BD Vacutainer® PST™ II tubes are supplied on request.
Frequently Bought Together