Order Entry
Denmark
ContactUsLinkComponent
 
3,5-Difluoroaniline 98%
 
undefined
3,5-Difluoroaniline 98%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to yellow Low melting solid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Melting point:
    37 to 41 °C
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together