2-Chloro-1,3-dinitrobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:606-21-3
:
Formula:C₆H₃ClN₂O₄
MDL Number:MFCD00014689
Boiling Pt:315 °C
Melting Pt:83…88 °C
Density:1.687
Molecular Weight:202.55 g/mol
Storage Temperature:Ambient
EINECS:210-107-6
ADR:6.1 II
Merck Index:13,02156
UN:3441
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together