3,5-Dichloroaniline 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:626-43-7
:
Formula:C₆H₅Cl₂N
MDL Number:MFCD00007774
Boiling Pt:260 °C (741 mmHg)
Melting Pt:49…53 °C
Density:1.58 g/cm³
Molecular Weight:162.02 g/mol
Flash Pt:133 °C
Storage Temperature:Ambient
EINECS:210-948-9
UN:3442
ADR:6.1 II
:GHS06
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to brown Crystals (May darken on storage)
- Infrared spectrum:Conforms
- Melting point:49 to 53 °C
- GC:≥97.5 %
- Fluorescence max:565 nm (in Methanol)
- Optimum lasing:588 nm (in Methanol)
- Molar absorption coefficient:106000 l/(mol.cm) (at Lambda max)
Specification Test Results
Frequently Bought Together