Water jet pump with non-return valve, DURAN®
181-9205
- Description:Water jet pump, flow rate 300 l/h, height 275 mm
- Environmentally Preferable:
- Pk:1
- Water pressure:Minimum 1,2 bar
DURAN®, borosilicate glass 3.3, clear.
With non-return valve.
DURAN®, borosilicate glass 3.3, clear.
With non-return valve.