Order Entry
Denmark
ContactUsLinkComponent
 
2-Methylpentane
Catalog # PROOCHE119
CAS Number:  
undefined
2-Methylpentane
Catalog # PROOCHE119
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Seq:
    2145
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 1,1-Dimethylbutane, Isohexane
 :
 :86.18 g/mol
EINECS:203-523-4
UN:1208
 :GHS02
 :GHS08
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:107-83-5
Formula:C₆H₁₄
MDL Number:MFCD00009406
Boiling Pt:62
Melting Pt:–154 °C
Density:0.653 g/cm³ (20 °C)
Flash Pt:–33 °C
ADR:3 II