Capric acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: Decylic acid, Decanoic acid
CAS Number:334-48-5
:
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00004441
Boiling Pt:268
Molecular Weight:172.27 g/mol
Flash Pt:147
Merck Index:14,01758
EINECS:206-376-4
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Solubility in EtOH (95vol%):almost transparency
Specification Test Results
Related Information
Frequently Bought Together