Diacetyl, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: 2,3-Butanedione
CAS Number:431-03-8
:
Formula:C₄H₆O₂
MDL Number:MFCD00008756
Boiling Pt:190
Melting Pt:–2 °C
Density:0.9904 g/cm³ (20 °C)
Molecular Weight:86.09 g/mol
Flash Pt:9
EINECS:207-069-8
UN:2346
ADR:3 II
Merck Index:13,02985
:M-Clarity™ quality level = MQ100
:GHS02
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together