Acetone, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
Boiling Pt:56.2
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Molecular Weight:58.08 g/mol
Flash Pt:<–20 °C (closed cup)
EINECS:200-662-2
UN:1090
ADR:3 II
Merck Index:13,00067
:M-Clarity™ quality level = MQ100
REACH:01-2119471330-49
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 C) soluble
- Melting Point:-95.4 C
- Molar Mass:58.08
- Boiling Point:56.2 C
- Vapor Pressure:233
- Flash Point:-18 C
- Refractive Index:1.35686
- Explosion Limit:2.6 - 12.8 %
- Density:0.79 g/cm3
- pH:5 - 6
- Ignition Point:465 C
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.00022.2500_TDS_EN ProductPage_100022
Product specification
msds_100022
SDS
Solvents for spectroscopy Uvasol
Product Literature
Frequently Bought Together