Order Entry
Denmark
ContactUsLinkComponent
Calcium chloride dihydrate 77 - 80% ≥77-80%, soluble, extra pure for drying
Calcium chloride dihydrate 77 - 80% ≥77-80%, soluble, extra pure for drying
Certificates
undefined
Calcium chloride dihydrate 77 - 80% ≥77-80%, soluble, extra pure for drying

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to light grey Flakes
  • Titration Complexometric:
    77 to 80 % (CaCl2)
  • Melting point:
    81 to 84 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together